CAS RN 41814-78-2
CAS RN 41814-78-2
IUPAC Name:
12-methyl-7-thia-2,4,5-triazatricyclo[6.4.0.0²,⁶]dodeca-1(12),3,5,8,10-pentaene
Synonyms:
tricyclazole
Molecular Weight (g/mol):
189.24
Molecular Formula:
C9H7N3S
InChi Key:
DQJCHOQLCLEDLL-UHFFFAOYSA-N
SMILES:
CC1=C2N3C=NN=C3SC2=CC=C1
1
–
2
of
2
results
1
Medchem Express Tricyclazole, MedChemExpress
MedChemExpress Tricyclazole is a pentaketide-derived melanin biosynthesis inhibitor and a unique fungicide for control of Pyricularia oryzae on rice.

2
SPEX Certiprep Beam, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material