CAS RN 41037-28-9
CAS RN 41037-28-9
IUPAC Name:
6-bromo-4-methylquinoline
Synonyms:
6-bromo-4-methylquinoline
Molecular Weight (g/mol):
222.09
Molecular Formula:
C10H8BrN
InChi Key:
XBZQSJLYRMZRSZ-UHFFFAOYSA-N
SMILES:
CC1=CC=NC2=CC=C(Br)C=C12
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 6-Bromo-4-methylquinoline, 96%
CAS: 41037-28-9 Molecular Formula: C10H8BrN Molecular Weight (g/mol): 222.085 MDL Number: MFCD09258893 InChI Key: XBZQSJLYRMZRSZ-UHFFFAOYSA-N Synonym: quinoline, 6-bromo-4-methyl,6-bromo-4-methyl-quinoline,6-bromo lepidine,4-methyl-6-bromoquinoline,quinoline,6-bromo-4-methyl,6-bromanyl-4-methyl-quinoline PubChem CID: 12475362 IUPAC Name: 6-bromo-4-methylquinoline SMILES: CC1=C2C=C(C=CC2=NC=C1)Br