accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 3999-55-1

CH3OO(R)(R)OOCH3

CAS RN 3999-55-1

IUPAC Name: 1,2-diethyl (1R,2R)-cyclopropane-1,2-dicarboxylate
Synonyms: 1,2-diethyl (1R,2R)-cyclopropane-1,2-dicarboxylate
Molecular Weight (g/mol): 186.21
Molecular Formula: C9H14O4
InChi Key: SXLDHZFJMXLFJU-RNFRBKRXSA-N
SMILES: CCOC(=O)[C@@H]1C[C@H]1C(=O)OCC
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.