accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 35150-09-5

CH3H3CCH3OOHNOH2N

CAS RN 35150-09-5

IUPAC Name: tert-butyl N-(carbamoylmethyl)carbamate
Synonyms: tert-butyl N-(carbamoylmethyl)carbamate
Molecular Weight (g/mol): 174.2
Molecular Formula: C7H14N2O3
InChi Key: RHONTQZNLFIDCQ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC(N)=O
Quantity 
  • (1)
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.