accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 33577-16-1

CH3SSCH3O

CAS RN 33577-16-1

IUPAC Name: methanesulfinyl(methylsulfanyl)methane
Synonyms: methyl methylthiomethyl sulphoxide
Molecular Weight (g/mol): 124.22
Molecular Formula: C3H8OS2
InChi Key: OTKFCIVOVKCFHR-UHFFFAOYNA-N
SMILES: CSCS(C)=O
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.