accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 306936-57-2

NHNOF

CAS RN 306936-57-2

IUPAC Name: 5-(4-fluorophenyl)-1H-pyrazole-4-carbaldehyde
Synonyms: 3-(4-fluorophenyl)-2H-pyrazole-4-carbaldehyde
Molecular Weight (g/mol): 190.18
Molecular Formula: C10H7FN2O
InChi Key: CMXTUUXWJBLVEH-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C1=C(C=O)C=NN1
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.