accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 27568-05-4

CH3OONOH3CCl

CAS RN 27568-05-4

IUPAC Name: ethyl 4-chloro-8-methoxyquinoline-3-carboxylate
Synonyms: ethyl 4-chloro-8-methoxyquinoline-3-carboxylate
Molecular Weight (g/mol): 265.69
Molecular Formula: C13H12ClNO3
InChi Key: BBOZDELEERNECG-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C2C(OC)=CC=CC2=C1Cl
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.