CAS RN 2600-69-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Phorate Oxon, TRC
CAS: 2600-69-3 Molekylær formel: C7 H17 O3 P S2 Molekylvægt (g/mol): 244.31 Synonym: Phosphorothioic acid, O,O-diethyl S-[(ethylthio)methyl] ester,Methanethiol, (ethylthio)-, S-ester with O,O-diethyl phosphorothioate (8CI),O,O-Diethyl S-(ethylthiomethyl) phosphorothioate,Phorate oxon,Phorate oxygen analog,Phorate thiolate analog,Phoratoxon,Thimet oxon,Thimet oxygen analog,Phorate-oxon IUPAC navn: 1-[ethoxy(ethylsulfanylmethylsulfanyl)fosforyl]oxyethan SMIL: CCOP(=O)(OCC)SCSCC