CAS RN 2549-51-1
IUPAC Navn:
ethenyl 2-chloroacetate
Synonymer:
ethenyl 2-chloroacetate
Molekylvægt (g/mol):
120.53
Molekylformel:
C4H5ClO2
InChi Key:
XJELOQYISYPGDX-UHFFFAOYSA-N
SMILES:
ClCC(=O)OC=C
1
–
1
af
1
Resultater
1
Vinylchloroacetat, 99%, stikk. med 4-methoxyphenol
CAS: 2549-51-1 Molekylær formel: C4H5ClO2 Molekylvægt (g/mol): 120.532 MDL nummer: MFCD00045275 InChI nøgle: XJELOQYISYPGDX-UHFFFAOYSA-N Synonym: vinyl chloroacetate,acetic acid, chloro-, ethenyl ester,acetic acid, 2-chloro-, ethenyl ester,acetic acid, chloro-, vinyl ester,chloroacetic acid vinyl ester,vinyl chloroacetate polymer,vinyl monochloroacetate,chloroacetic acid vinyl,acetic acid, chloro-, vinyl ester, polymers,acmc-1clv7 PubChem CID: 82156 IUPAC navn: ethenyl-2-chloracetat SMIL: C=COC(=O)CCl