CAS RN 2499-59-4
CAS RN 2499-59-4
IUPAC Navn:
octyl prop-2-enoate
Synonymer:
octyl acrylate
Molekylvægt (g/mol):
184.28
Molekylformel:
C11H20O2
InChi Key:
ANISOHQJBAQUQP-UHFFFAOYSA-N
SMILES:
CCCCCCCCOC(=O)C=C
1
–
2
af
2
Resultater
1
n-Octyl Acrylate (Stabilized with MEHQ), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
n-Octyl Acrylate (Stabilized with MEHQ), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.