CAS RN 2498-46-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
2-Phenylacetimidamid Hydrochlorid, TRC
CAS: 2498-46-6 Molekylær formel: C8H10N2 . HCl Molekylvægt (g/mol): 170.64 Synonym: Benzeneethanimidamide, hydrochloride (1:1) (ACI),Acetamidine, 2-phenyl-, hydrochloride (6CI, 7CI),Acetamidine, 2-phenyl-, monohydrochloride (8CI),Benzeneethanimidamide, monohydrochloride (9CI),2-Phenylacetamidine hydrochloride,Phenylacetamidine hydrochloride,α-Phenylacetamidine hydrochloride,2-Phenylacetimidamide hydrochloride IUPAC navn: 2-phenylethanimidamid; hydrochlorid SMIL: Cl.NC(=N)Cc1ccccc1