CAS RN 2498-27-3
CAS RN 2498-27-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
3-Hydroxyphenyltrimethylammonium Iodide, TRC
CAS: 2498-27-3 Molekylær formel: C9H14INO Molekylvægt (g/mol): 279.12 Synonym: 3-Hydroxy-N,N,N-trimethylbenzenaminium Iodide,(m-Hydroxyphenyl)trimethylammonium Iodide,3-(Trimethylammonio)phenol Iodide,Trimethyl(3-hydroxyphenyl)ammonium Iodide,m-(Dimethylamino)phenol Methiodide,m-Hydroxy-N-trimethylanilinium Iodide,m-Hydroxytrimethylanilinium Iodide IUPAC navn: (3-hydroxyphenyl)-trimethylazanium;iodide SMIL: C[N+](C)(C)C1=CC(=CC=C1)O.[I-]