CAS RN 24938-37-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
207.2
Molekylformel:
C8H15O6
InChi Key:
CODQTHDBFINLJQ-UHFFFAOYSA-N
SMILES:
[O]CCO.[O][C](O)CCCCC(O)=O
1
–
1
af
1
Resultater
1
Poly(Ethylen Adipate)~ 10.000 Ifølge GPC, TRC
CAS: 24938-37-2 Molekylær formel: C6H10O4 . C2H6O2 Synonym: 1,2-Ethanediol-adipic Acid Copolymer,Adipic Acid-1,2-ethanediol Copolymer,Adipic Acid-ethylene Glycol Copolymer,Adipic Acid-ethylene Glycol Polyester,Adipic Acid-ethylene Glycol Polymer,Hexanedioic acid-ethylene glycol copolymer,PEGA,Poly(Ethylene Glycol Adipate),Ethylene Glycol-adipic Acid Polymer IUPAC navn: bis(2-hydroxyethyl) hexanedioat SMIL: OCCOC(=O)CCCCC(=O)OCCO