CAS RN 2492423-29-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
R&D Systems™ Molnupiravir
A Tocris product, part of R&D Systems. Prodrug of antiviral nucleoside analog; inhibits SARS-CoV, SARS-CoV-2 and MERS-CoV infection
2
EIDD 2801, TRC
CAS: 2492423-29-5 Molekylær formel: C13 H19 N3 O7 Molekylvægt (g/mol): 329.306 Synonym: Uridine, 4-oxime, 5'-(2-methylpropanoate), (4Z)- (ACI),EIDD 1931-isopropyl ester,EIDD 2801,Lagevrio,MK-4482,Molnupiravir,β-D-N4 hydroxycytidine-5'-isopropyl ester,Uridine (4Z)-4-oxime 5'-(2-methylpropanoate) IUPAC navn: [(2R,3S,4R,5R)-3,4-dihydroxy-5-[(4Z)-4-hydroxyimino-2-oxopyrimidin-1-yl]oxolan-2-yl]methyl2-methylpropanoat SMIL: CC(C)C(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=C\C(=N\O)\NC2=O