CAS RN 2484-30-2
CAS RN 2484-30-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
17beta-Hydroxyandrosta-4,6-dien-3-one (Δ6-Testosterone), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
∆6-Testosterone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
∆6-Testosterone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.