CAS RN 2481-09-6
IUPAC Navn:
hydrogen (2R)-2-amino-3-(tert-butylsulfanyl)propanoic acid chloride
Synonymer:
hydrogen (2R)-2-amino-3-(tert-butylsulfanyl)propanoic acid chloride
Molekylvægt (g/mol):
213.72
Molekylformel:
C7H16ClNO2S
InChi Key:
MHBMYFJKEBCMDR-JEDNCBNOSA-N
SMILES:
[H+].[Cl-].CC(C)(C)SC[C@H](N)C(O)=O
1
–
1
af
1
Resultater
1
S-tert-Butyl-L-cysteine hydrochloride, 98%
CAS: 2481-09-6 MDL nummer: MFCD00037426