CAS RN 247565-03-3
CAS RN 247565-03-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(4S)-6-Chloro-3,4-dihydro-4-hydroxy-3-[(1R)-1-phenylethyl]-4-(trifluoromethyl)-2(1H)quinazolinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(4S)-6-Chloro-3,4-dihydro-4-hydroxy-3-[(1R)-1-phenylethyl]-4-(trifluoromethyl)-2(1H)quinazolinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(4S)-6-Chloro-3,4-dihydro-4-hydroxy-3-[(1R)-1-phenylethyl]-4-(trifluoromethyl)-2(1H)quinazolinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.