CAS RN 2475-31-2
CAS RN 2475-31-2
IUPAC Navn:
(Z)-5,5',7,7'-tetrabromo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-3,3'-dione
Synonymer:
C.I. vat blue 5
Molekylvægt (g/mol):
577.85
Molekylformel:
C16H6Br4N2O2
InChi Key:
PTWYQANXSNMUTI-YPKPFQOOSA-N
SMILES:
BrC1=CC(Br)=C2N\C(C(=O)C2=C1)=C1/NC2=C(Br)C=C(Br)C=C2C1=O
1
–
3
af
3
Resultater
1
Ciba Blue 2B (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Ciba Blue 2B (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Ciba Blue 2B (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.