CAS RN 2472-88-0
CAS RN 2472-88-0
IUPAC Navn:
bis(tetrabutylazanium) sulfate
Synonymer:
bis(tetrabutylammonium ion) sulfate
Molekylvægt (g/mol):
581
Molekylformel:
C32H72N2O4S
InChi Key:
ZXUCBXRTRRIBSO-UHFFFAOYSA-L
SMILES:
[O-]S([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC
1
–
1
af
1
Resultater
1
Tetra-n-butylammonium sulfate, 50% w/w aq. soln.
CAS: 2472-88-0 Molekylær formel: C32H72N2O4S Molekylvægt (g/mol): 581.00 MDL nummer: MFCD00216632 InChI nøgle: ZXUCBXRTRRIBSO-UHFFFAOYSA-L Synonym: bis tetrabutylammonium sulphate,tetrabutylammonium sulfate,tetrabutylammonium sulfate solution,1-butanaminium, n,n,n-tributyl-, sulfate 2:1,tetrabutyl ammoniumsulfat,tetrabutylammonium sulphate,acmc-1cdo3,tetrabutyl ammonium sulfate,tetrabutyl-ammonium sulfate,tetra-n-butylammonium sulfate PubChem CID: 11433187 IUPAC navn: tetrabutylazanium;sulfat SMIL: [O-]S([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC