CAS RN 247068-85-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(2S)-2-Amino-4-methyl-1-[(2R)-2-methyloxiranyl]-1-pentanone Trifluoroacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(2S)-2-Amino-4-methyl-1-[(2R)-2-methyloxiranyl]-1-pentanone Trifluoroacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(2S)-2-Amino-4-methyl-1-[(2R)-2-methyloxiranyl]-1-pentanone Trifluoroacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.