CAS RN 24679-92-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Fluoro 2-Deoxy-2-fluoro-3,4,6-tri-O-acetyl-D-mannopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Fluoro 2-Deoxy-2-fluoro-3,4,6-tri-O-acetyl-D-mannopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.