CAS RN 2462-32-0
CAS RN 2462-32-0
IUPAC Navn:
benzyl 2-amino-3-phenylpropanoate hydrochloride
Synonymer:
benzyl 2-amino-3-phenylpropanoate hydrochloride
Molekylvægt (g/mol):
291.78
Molekylformel:
C16H18ClNO2
InChi Key:
CEXFHIYDTRNBJD-UHFFFAOYNA-N
SMILES:
Cl.NC(CC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1
1
–
2
af
2
Resultater
1
L-Phenylalanine benzyl ester hydrochloride, 98%
CAS: 2462-32-0 Molekylær formel: C16H18ClNO2 Molekylvægt (g/mol): 291.78 MDL nummer: MFCD00043249 InChI nøgle: CEXFHIYDTRNBJD-RSAXXLAASA-N Synonym: l-phenylalanine benzyl ester hydrochloride,h-phe-obzl.hcl,h-phe-obzl hcl,l-phenylalanine benzyl ester hcl,s-benzyl 2-amino-3-phenylpropanoate hydrochloride,phenylalanine, phenylmethyl ester, hydrochloride,benzyl 2s-2-amino-3-phenylpropanoate hydrochloride,phenylmethyl 2s-2-amino-3-phenylpropanoate, chloride,pubchem19007,benzyl phenylalaninate hydrochloride PubChem CID: 12636208 IUPAC navn: benzyl(2S)-2-amino-3-phenylpropanoat;hydrochlorid SMIL: Cl.N[C@@H](CC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1