CAS RN 246021-75-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
2-[2-(2-Amino-9H-purin-9-yl)ethyl]propane-1,3-diol Hydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
6-Deoxypenciclovir hydrochloride, TRC
CAS: 246021-75-0 Molekylær formel: C10 H15 N5 O2 . Cl H Molekylvægt (g/mol): 273.72 Synonym: 1,3-Propanediol, 2-[2-(2-amino-9H-purin-9-yl)ethyl]-, hydrochloride (1:1),1,3-Propanediol, 2-[2-(2-amino-9H-purin-9-yl)ethyl]-, monohydrochloride (9CI) IUPAC navn: 2-[2-(2-aminopurin-9-yl)ethyl]propane-1,3-diol;hydrochloride SMIL: Cl.Nc1ncc2ncn(CCC(CO)CO)c2n1