CAS RN 245342-14-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
LED209, MedChemExpress
MedChemExpress LED209 is a potent small molecule inhibitor of bacterial receptor QseC, is a potent prodrug that is highly selective for QseC.