CAS RN 24517-64-4
CAS RN 24517-64-4
IUPAC Navn:
2-amino-3-cyanopyridin-1-ium
Synonymer:
2-amino-3-cyanopyridin-1-ium
Molekylvægt (g/mol):
120.13
Molekylformel:
C6H6N3
InChi Key:
YYXDQRRDNPRJFL-UHFFFAOYSA-O
SMILES:
NC1=[NH+]C=CC=C1C#N
1
–
3
af
3
Resultater
1
2-amino-3-cyanopyridin, 98 %, Thermo Scientific Chemicals
CAS: 24517-64-4 Molekylær formel: C6H6N3 Molekylvægt (g/mol): 120.13 MDL nummer: MFCD00128867 InChI nøgle: YYXDQRRDNPRJFL-UHFFFAOYSA-O Synonym: 2-amino-3-cyanopyridine,2-aminonicotinonitrile,2-amino-3-pyridinecarbonitrile,2-amino-pyridine-3-carbonitrile,2-amino-nicotinonitrile,3-pyridinecarbonitrile, 2-amino,3-cyanopyridin-2-amine,3-cyanopyridine-2-amine,aminonicotinonitrile,zlchem 506 PubChem CID: 818259 IUPAC navn: 2-amino-3-cyanopyridin-1-ium SMIL: NC1=[NH+]C=CC=C1C#N
2
2-Amino-3-pyridinecarbonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.