CAS RN 219911-35-0
CAS RN 219911-35-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Tocris Bioscience™ MPEP hydrochloride
Potent mGlu5 antagonist; also positive allosteric modulator of mGlu4 receptors
2
Medchem Express MPEP Hydrochloride, MedChemExpress
MedChemExpress MPEP Hydrochloride is a potent, selective, noncompetitive, orally active and systemically active mGlu5 receptor antagonist, with an IC50 of 36 nM for completely inhibiting quisqualate-stimulated phosphoinositide (PI) hydrolysis. MPEP Hydrochloride has anxiolytic-or antidepressant-like effects.

3
Tocris Bioscience™ MPEP hydrochloride
Potent mGlu5 antagonist; also positive allosteric modulator of mGlu4 receptors

4
Tocris Bioscience™ MPEP hydrochloride
Potent mGlu5 antagonist; also positive allosteric modulator of mGlu4 receptors
