Få mere at vide
Tocris Bioscience™ MPEP hydrochloride
Potent mGlu5 antagonist; also positive allosteric modulator of mGlu4 receptors
Brand: Tocris Bioscience™ 1212/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
MPEP hydrochloride is a potent and highly selective non-competitive antagonist at the mGlu5 receptor subtype (IC50 = 36 nM) and a positive allosteric modulator at mGlu4 receptors. Centrally active following systemic administration in vivo. Reverses mechanical hyperalgesia in the inflamed rat hind paw.Tekniske data
| 2-Methyl-6-(phenylethynyl)pyridine hydrochloride | |
| >99% | |
| Soluble to 100mM in DMSO |
| 219911-35-0 | |
| 50 mg |
Produkttitel
Ser du en mulighed for forbedring?
For Research Use Only