CAS RN 2159-18-4
CAS RN 2159-18-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
1-Hexanol-d11, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
n-Hexyl-2,2,3,3,4,4,5,5,6,6,6-d11 Alcohol, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
3
1-Hexanol-d11, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
1-Hexanol-d11, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.