Få mere at vide
1-Hexanol-d11, TRC
Artikelnummer. 30406938
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30406938 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30406938 Leverandør Toronto Research Chemicals Leverandørnr. TRCH281222500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Hexanol-2,2,3,3,4,4,5,5,6,6,6-D11
CAS 2159-18-4
Molekylær formel C6 D11 H3 O
Mængde 500 mg
Synonym 1-Hexanol-d11, 1-Hexan-2,2,3,3,4,4,5,5,6,6,6-d11-ol (ACI), Hexyl-2,2,3,3,4,4,5,5,6,6,6-d11 alcohol (7CI, 8CI), 1-Hexanol-d11, 1-Hexanol-2,2,3,3,4,4,5,5,6,6,6-D11, n-Hexyl-2,2,3,3,4,4,5,5,6,6,6-d11 alcohol, 1-Hexanol-2,2,3,3,4,4,5,5,6,6,6-d11
InChI formel InChI=1S/C6H14O/c1-2-3-4-5-6-7/h7H,2-6H2,1H3/i1D3,2D2,3D2,4D2,5D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])CO
IUPAC navn 2,2,3,3,4,4,5,5,6,6,6-undecadeuteriohexan-1-ol
Molekylvægt (g/mol) 113.243
Formel vægt 113.174