CAS RN 2152628-33-4
CAS RN 2152628-33-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ Selpercatinib
Potent and selective RET inhibitor
2
Medchem Express Selpercatinib, MedChemExpress
MedChemExpress Selpercatinib (LOXO-292) is a potent, selective RET kinase inhibitor with IC50 values of 14.0 nM, 24.1 nM, and 530.7 nM for RET (WT), RET (V804M), and RET (G810R), respectively. Selpercatinib has anticancer activity.
