accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 20427-58-1

H2OZnOH2

CAS RN 20427-58-1

IUPAC Name: zinc hydroxide
Synonyms: zinc hydroxide
Molecular Weight (g/mol): 101.41
Molecular Formula: H4O2Zn
InChi Key: JQBMTMBJMXRRCJ-UHFFFAOYSA-N
SMILES: O.O.[Zn]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.