accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 196868-63-0

H3CNNHONHClOCH3

CAS RN 196868-63-0

IUPAC Name: 1-(5-chloro-2-methoxyphenyl)-3-(2-methylquinolin-4-yl)urea
Synonyms: 1-(5-chloro-2-methoxyphenyl)-3-(2-methylquinolin-4-yl)urea
Molecular Weight (g/mol): 341.8
Molecular Formula: C18H16ClN3O2
InChi Key: YBLWOZUPHDKFOT-UHFFFAOYSA-N
SMILES: COC1=C(NC(=O)NC2=C3C=CC=CC3=NC(C)=C2)C=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.