CAS RN 19458-49-2
CAS RN 19458-49-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
17,20:20,21-Bis(methylenedioxy)-5b-pregnan-3b-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
17,20:20,21-Bis(methylenedioxy)-5Beta-pregnan-3Beta-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
17,20:20,21-Bis(methylenedioxy)-5Beta-pregnan-3Beta-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.