CAS RN 19130-96-2
CAS RN 19130-96-2
IUPAC Navn:
(2R,3R,4R,5S)-2-(hydroxymethyl)piperidine-3,4,5-triol
Synonymer:
1-deoxynojirimycin
Molekylvægt (g/mol):
163.17
Molekylformel:
C6H13NO4
InChi Key:
LXBIFEVIBLOUGU-JGWLITMVSA-N
SMILES:
OC[C@H]1NC[C@H](O)[C@@H](O)[C@@H]1O
1
–
2
af
2
Resultater
2
1-Deoxynojirimycin, MedChemExpress
MedChemExpress 1-Deoxynojirimycin (Duvoglustat) is a potent and orally active α-glucosidase inhibitor. 1-Deoxynojirimycin suppresses postprandial blood glucose and is widely used for diabetes mellitus. 1-Deoxynojirimycin possesses antihyperglycemic, anti-obesity, and antiviral features.
