CAS RN 1819363-80-8
CAS RN 1819363-80-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ PFI 3
Potent and selective SMARCA2/4 and polybromo 1 inhibitor
2
Medchem Express PFI-3, MedChemExpress
MedChemExpress PFI-3 is a selective, potent and cell-permeable SMARCA2/4 bromodomain inhibitor with a Kd of 89 nM.
