Få mere at vide
Tocris Bioscience™ PFI 3
Beskrivelse
PFI 3 is a potent and selective polybromo 1 (PBRM1) and SMARCA4 inhibitor (Kd values are 48 and 89 nM respectively) that also inhibits SMARCA2. This compoud displays 30-fold selectivity over other sub-family branches. PFI 3 accelerates FRAP recovery in cells at a concentration of 1 μM.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (2E)-1-(2-Hydroxyphenyl)-3-[(1R,4R)-5-(pyridin-2-yl)-2,5-diazabicyclo[2.2.1]heptan-2-yl]prop-2-en-1-one |
| CAS | 1819363-80-8 |
| Mængde | 10 mg |
| Molekylær formel | C19H19N3O2 |
| Formel vægt | 321.37 |
| Renhed | 98% |