CAS RN 175277-40-4
CAS RN 175277-40-4
IUPAC Name:
1-(1H-pyrazol-5-yl)ethan-1-one hydrochloride
Synonyms:
1-(2H-pyrazol-3-yl)ethanone hydrochloride
Molecular Weight (g/mol):
146.57
Molecular Formula:
C5H7ClN2O
InChi Key:
MTFWBEDWXAHBNR-UHFFFAOYSA-N
SMILES:
Cl.CC(=O)C1=CC=NN1
1
–
1
of
1
results
1
Thermo Scientific Maybridge 1-(1H-Pyrazol-5-yl)ethan-1-one hydrochloride, ≥95%, Thermo Scientific™
CAS: 175277-40-4 Molecular Formula: C5H7ClN2O Molecular Weight (g/mol): 146.574 MDL Number: MFCD00204160 InChI Key: MTFWBEDWXAHBNR-UHFFFAOYSA-N Synonym: 1-1h-pyrazol-5-yl ethan-1-one hydrochloride,1-1h-pyrazol-5-yl ethanone hydrochloride,1-1h-pyrazol-3-yl ethanone hydrochloride,1-2h-pyrazol-3-yl ethanone hydrochloride,5-acetyl-1h-pyrazole hydrochloride,5-acetylpyrazole, chloride,3-acetylpyrazole hydrochloride,5-acetylpyrazole hydrochloride,1-1h-pyrazol-5-yl-1-ethanone hydrochloride,1-1h-pyrazole-5-yl ethan-1-one hydrochloride PubChem CID: 2799623 IUPAC Name: 1-(1H-pyrazol-5-yl)ethanone;hydrochloride SMILES: CC(=O)C1=CC=NN1.Cl