CAS RN 175135-32-7
CAS RN 175135-32-7
IUPAC Name:
4-(3,4-difluorophenyl)-1,3-thiazol-2-amine
Synonyms:
4-(3,4-difluorophenyl)-1,3-thiazol-2-amine
Molecular Weight (g/mol):
212.22
Molecular Formula:
C9H6F2N2S
InChi Key:
NDCSJUJQMRFHEX-UHFFFAOYSA-N
SMILES:
NC1=NC(=CS1)C1=CC(F)=C(F)C=C1
1
–
1
of
1
results
1
Thermo Scientific Maybridge 4-(3,4-Difluorophenyl)-1,3-thiazol-2-amine, 97%, Thermo Scientific™
CAS: 175135-32-7 Molecular Formula: C9H6F2N2S Molecular Weight (g/mol): 212.218 MDL Number: MFCD00052876 InChI Key: NDCSJUJQMRFHEX-UHFFFAOYSA-N Synonym: 4-3,4-difluorophenyl-1,3-thiazol-2-amine,2-amino-4-3,4-difluorophenyl thiazole,4-3,4-difluorophenyl thiazol-2-amine,4-3,4-difluoro-phenyl-thiazol-2-ylamine,2-amino-4-3,4-difluorophenyl-1,3-thiazole,2-thiazolamine,4-3,4-difluorophenyl,4-3,4-difluoro-phenyl-1,3-thiazol-2-amine,4-3,4-difluorophenyl-1,3-thiazole-2-ylamine,pubchem14456,timtec-bb sbb005523 PubChem CID: 706326 IUPAC Name: 4-(3,4-difluorophenyl)-1,3-thiazol-2-amine SMILES: C1=CC(=C(C=C1C2=CSC(=N2)N)F)F