accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 157021-61-9

ClNClFFF

CAS RN 157021-61-9

IUPAC Name: 2,6-dichloro-4-(trifluoromethyl)benzonitrile
Synonyms: 2,6-dichloro-4-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 240.01
Molecular Formula: C8H2Cl2F3N
InChi Key: NCXSSFQXQAOREM-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=C(C#N)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.