CAS RN 155815-98-8
CAS RN 155815-98-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MedChemExpress Hoechst 33342 analog 2 trihydrochloride
MedChemExpress Hoechst 33342 analog 2 trihydrochloride is a anglog of Hoechst 33342. Hoechst 33342 is a DNA minor groove binder used fluorochrome for visualizing cellular DNA.
