accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 154-92-7

H2NNH2NNHOOHO

CAS RN 154-92-7

IUPAC Name: 5-[(diaminomethylidene)amino]-2-(phenylformamido)pentanoic acid
Synonyms: 5-[(diaminomethylidene)amino]-2-(phenylformamido)pentanoic acid
Molecular Weight (g/mol): 278.31
Molecular Formula: C13H18N4O3
InChi Key: RSYYQCDERUOEFI-UHFFFAOYNA-N
SMILES: NC(N)=NCCCC(NC(=O)C1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.