accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 141580-71-4

OON

CAS RN 141580-71-4

IUPAC Name: 2-(pyridin-2-yloxy)benzaldehyde
Synonyms: 2-(pyridin-2-yloxy)benzaldehyde
Molecular Weight (g/mol): 199.21
Molecular Formula: C12H9NO2
InChi Key: QCKZHTQRJYCZTD-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC=C1OC1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.