accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1393814-38-4

CH3ONNOOHCl

CAS RN 1393814-38-4

IUPAC Name: 5-(3-chloro-4-cyclohexylphenyl)-1-(3-methoxyphenyl)-1H-pyrazole-3-carboxylic acid
Synonyms: 5-(3-chloro-4-cyclohexylphenyl)-1-(3-methoxyphenyl)pyrazole-3-carboxylic acid
Molecular Weight (g/mol): 410.9
Molecular Formula: C23H23ClN2O3
InChi Key: BNALUYKEGYUHQC-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)N1N=C(C=C1C1=CC(Cl)=C(C=C1)C1CCCCC1)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.