CAS RN 131-72-6
CAS RN 131-72-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Medchem Express Meptyldinocap, MedChemExpress
MedChemExpress Meptyldinocap (2,4-DNOPC) is a novel powdery mildew (Erysiphe necator) fungicide which shows protectant and post-infective activities.
