CAS RN 13009-99-9
CAS RN 13009-99-9
IUPAC Name:
4-(aminomethyl)benzene-1-sulfonamide; acetic acid
Synonyms:
acetic acid; emilene
Molecular Weight (g/mol):
246.28
Molecular Formula:
C9H14N2O4S
InChi Key:
UILOTUUZKGTYFQ-UHFFFAOYSA-N
SMILES:
CC(O)=O.NCC1=CC=C(C=C1)S(N)(=O)=O
1
–
1
of
1
results
1
Medchem Express Mafenide Acetate, MedChemExpress
MedChemExpress Mafenide Acetate is an effective sulfonamide-type antimicrobial agent used for burn wounds. Mafenide Acetate shows activity against both Gram-positive and Gram-negative organisms, including Pseudomonas aeruginosa, via inhibition of nucleotide synthesis.
