CAS RN 1018673-42-1
CAS RN 1018673-42-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express HDAC-IN-3, MedChemExpress
MedChemExpress HDAC-IN-3 is a histone deacetylase (HDAC) inhibitor, extracted from patent WO/2008040934 A1.
