CAS RN 998-07-2
IUPAC Navn:
(2-aminoethoxy)[2,3-bis(tetradecanoyloxy)propoxy]phosphinic acid
Synonymer:
DMPE dimyristoylcephaline 1,2-dimyristoyl-3-cephalin cephalin, 1,2-dimyristoyl- dimyristoylcephalin dimyristoylphosphatidylethanolamine 1,2-dimyristoyl-cephalin
Molekylvægt (g/mol):
635.86
Molekylformel:
C33H66NO8P
InChi Key:
NEZDNQCXEZDCBI-UHFFFAOYNA-N
SMILES:
CCCCCCCCCCCCCC(=O)OCC(COP(O)(=O)OCCN)OC(=O)CCCCCCCCCCCCC
1
–
1
af
1
Resultater
1
1,2-Dimyristoyl-sn-glycero-phosphoethanolamine (DMPE), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.