Få mere at vide
1,2-Dimyristoyl-sn-glycero-phosphoethanolamine (DMPE), Mikromol™
Beskrivelse
1,2-Dimyristoyl-sn-glycero-phosphoethanolamine (DMPE)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 1,2-Dimyristoyl-sn-glycero-phosphoethanolamine |
| CAS | 998-07-2 |
| Analyt- eller komponentnavne | 1,2-Dimyristoyl-sn-glycero-phosphoethanolamine |
| Forsendelsestilstand | Ice pack (-20°C) |
| Molekylær formel | C33 H66 N O8 P |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Tetradecanoic acid, (1R)-1-[[[(2-aminoethoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl ester, Myristin, 1,2-di-, 2-aminoethyl hydrogen phosphate, L- (8CI), Tetradecanoic acid, 1-[[[(2-aminoethoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl ester, (R)-, Phosphoric acid, mono(2-aminoethyl) ester, monoester with 1,2-dimyristin, L- (8CI), (R)-Dimyristoylphosphatidylethanolamine, 1,2-Dimyristoyl-3-sn-phosphatidylethanolamine, 1,2-Dimyristoyl-L-3-phosphatidylethanolamine, 1,2-Dimyristoyl-sn-glycero-3-phosphatidylethanolamine, 1,2-Dimyristoyl-sn-glycero-3-phosphoethanolamine, 1,2-Dimyristoyl-sn-glycero-3-phosphoethanolamine, 1,2-Dimyristoyl-sn-glycero-3-phosphorylethanolamine, 1,2-Dimyristoyl-sn-glycero-phosphoethanolamine, 1,2-Dimyristoylsn-glycero-3-phospholethanolamine, 1,2-Ditetradecanoyl-sn-glycero-3-phosphoethanolamine, DMPE, Dimyristoyl-L-α-phosphatidylethanolamine, Dimyristoyl-sn-glycerol-3-phosphorylethanolamine, L-α-Dimyristoylphosphatidylethanolamine, L-β,γ-Dimyristoyl-α-phosphatidylethanolamine, β,γ-Dimyristoyl-L-α-phosphatidylethanolamine |
| InChI formel | InChI=1S/C33H66NO8P/c1-3-5-7-9-11-13-15-17-19-21-23-25-32(35)39-29-31(30-41-43(37,38)40-28-27-34)42-33(36)26-24-22-20-18-16-14-12-10-8-6-4-2/h31H,3-30,34H2,1-2H3,(H,37,38)/t31-/m1/s1 |
| SMIL | CCCCCCCCCCCCCC(=O)OC[C@H](COP(=O)(O)OCCN)OC(=O)CCCCCCCCCCCCC |
| IUPAC navn | [(2R)-3-[2-aminoethoxy(hydroxy)phosphoryl]oxy-2-tetradecanoyloxypropyl] tetradecanoate |
| Vis mere |