CAS RN 98614-76-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Tocris Bioscience™ Parathyroid hormone (1-34) (rat)
Parathyroid hormone (PTH) receptor agonist
3