Få mere at vide
Tocris Bioscience™ Parathyroid hormone (1-34) (rat)
Beskrivelse
Parathyroid hormone (1-34) (rat) is a parathyroid hormone (PTH) receptor agonist. Increases serum PTH levels and bone mass in rats.
Tekniske data
Tekniske data
| Arter | Rat |
| Renhed | 95% |
| CAS | 98614-76-7 |
| Indhold og opbevaring | Store at −20°C |
| Til brug med (applikation) | Biological reactions |
| Molekylvægt (g/mol) | 4057.74g/mol |
| Produkttype | Protein agonist |
| Mængde | 1 mg |
| Regulatorisk status | RUO - research use only |
| Sekvens | AVSEIQLMHNLGKHLASVERMQWLRKKLQDVHNF |
For Research Use Only